CAS RN 27262-48-2
CAS RN 27262-48-2
IUPAC Name:
hydrogen (2S)-1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide chloride
Synonyms:
hydrogen levobupivacaine chloride
Molecular Weight (g/mol):
324.89
Molecular Formula:
C18H29ClN2O
InChi Key:
SIEYLFHKZGLBNX-NTISSMGPSA-N
SMILES:
[H+].[Cl-].CCCCN1CCCC[C@H]1C(=O)NC1=C(C)C=CC=C1C
1
–
1
of
1
results
1
Thermo Scientific™ Levobupivacaine hydrochloride, 98%
CAS: 27262-48-2 Molecular Formula: C18H29ClN2O Molecular Weight (g/mol): 324.89 MDL Number: MFCD01704265 InChI Key: SIEYLFHKZGLBNX-NTISSMGPSA-N IUPAC Name: hydrogen (2S)-1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide chloride SMILES: [H+].[Cl-].CCCCN1CCCC[C@H]1C(=O)NC1=C(C)C=CC=C1C