accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27361-16-6

IUPAC Name: 2-butoxypyridine
Synonyms: 2-butoxypyridine
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: OFLSKXBALZCMCX-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results