Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butoxypyridine 97.0+%, TCI America™
SDP

Catalog No. B192525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B192525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B192525G Supplier TCI America Supplier No. B192525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27361-16-6
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00006267
InChI Key OFLSKXBALZCMCX-UHFFFAOYSA-N
Synonym 2-n-butoxypyridine, butyl 2-pyridyl ether, pyridine, 2-butoxy, acmc-209gwm, 2-butoxypyridine, 2-butoxypyridine, tech.
PubChem CID 95701
IUPAC Name 2-butoxypyridine
SMILES CCCCOC1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 200°C
Quantity 25 g
Formula Weight 151.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Butoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.