accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2738-18-3

IUPAC Name: (3E)-2,6-dimethylhept-3-ene
Synonyms: (3E)-2,6-dimethylhept-3-ene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: KDISTZUHDQPXDE-AATRIKPKSA-N
SMILES: CC(C)C\C=C\C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results