accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27431-40-9

IUPAC Name: 3-methoxybutyl 3-sulfanylpropanoate
Synonyms: 3-methoxybutyl 3-sulfanylpropanoate
Molecular Weight (g/mol): 192.27
Molecular Formula: C8H16O3S
InChi Key: APWBGRBFKMJPLW-UHFFFAOYNA-N
SMILES: COC(C)CCOC(=O)CCS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results