Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Methoxybutyl 3-Mercaptopropionate 97.0+%, TCI America™
SDP

Catalog No. M1802500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1802500G 500 g
M180225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1802500G Supplier TCI America Supplier No. M1802500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 27431-40-9
Molecular Formula C8H16O3S
Molecular Weight (g/mol) 192.273
MDL Number MFCD04038137
InChI Key APWBGRBFKMJPLW-UHFFFAOYSA-N
Synonym 3-Mercaptopropionic Acid 3-Methoxybutyl Ester
PubChem CID 122398
IUPAC Name 3-methoxybutyl 3-sulfanylpropanoate
SMILES CC(CCOC(=O)CCS)OC

Specifications

Color Yellow
Quantity 500 g
Formula Weight 192.27
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methoxybutyl 3-Mercaptopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.