accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHOH

CAS RN 27489-60-7

IUPAC Name: N-(4-hydroxycyclohexyl)acetamide
Synonyms: N-(4-hydroxycyclohexyl)acetamide
Molecular Weight (g/mol): 157.21
Molecular Formula: C8H15NO2
InChi Key: HWAFCRWGGRVEQL-UHFFFAOYSA-N
SMILES: CC(=O)NC1CCC(O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results