accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2752-17-2

IUPAC Name: 2-(2-aminoethoxy)ethan-1-amine
Synonyms: 2-(2-aminoethoxy)ethanamine
Molecular Weight (g/mol): 104.15
Molecular Formula: C4H12N2O
InChi Key: GXVUZYLYWKWJIM-UHFFFAOYSA-N
SMILES: NCCOCCN
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)