Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Oxybis(ethylamine), 98%, Thermo Scientific™

Catalog No. AC275840025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC275840025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC275840025 Supplier Thermo Scientific Chemicals Supplier No. 275840025

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2752-17-2
Molecular Formula C4H12N2O
Molecular Weight (g/mol) 104.15
MDL Number MFCD00466591
InChI Key GXVUZYLYWKWJIM-UHFFFAOYSA-N
Synonym 2,2'-oxydiethanamine, 1,5-diamino-3-oxapentane, 2,2'-oxybis ethylamine, 2-2-aminoethoxy ethanamine, ethanamine, 2,2'-oxybis, bis 2-aminoethyl ether, aminoethyl ether, 2,2'-diaminodiethyl ether, unii-ffw1773zz6, 2,2'-oxydiethylamine
PubChem CID 75982
IUPAC Name 2-(2-aminoethoxy)ethan-1-amine
SMILES NCCOCCN

Specifications

Density 0.9610g/mL
Boiling Point 69.0°C to 71.0°C (15.0 mmHg)
Refractive Index 1.4550 to 1.4580
Linear Formula NH2CH2CH2OCH2CH2NH2
Quantity 2.5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform
Formula Weight 104.15
Chemical Name or Material 2,2'-Oxybis(ethylamine)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.