accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2752-99-0

IUPAC Name: 1,3-bis[(13Z)-docos-13-enoyloxy]propan-2-yl (13Z)-docos-13-enoate
Synonyms: trierucin
Molecular Weight (g/mol): 1053.78
Molecular Formula: C69H128O6
InChi Key: XDSPGKDYYRNYJI-IUPFWZBJSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC\C=C/CCCCCCCC)OC(=O)CCCCCCCCCCC\C=C/CCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results