CAS RN 27536-56-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ 4α-Phorbol-12,13-didecanoate
Inactive on mouse skin as an inflammatory and tumor-promoting agent.