CAS RN 2761281-50-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ PROTAC™(H-PGDS)-7
Potent hematopoietic prostaglandin D2 synthase Degrader (PROTAC™)