CAS RN 276695-22-8
IUPAC Name:
8,9-dichloro-1H,2H,3H,4H,4aH,5H,6H-pyrazino[1,2-a]quinoxalin-5-one hydrochloride
Synonyms:
8,9-dichloro-1H,2H,3H,4H,4aH,6H-pyrazino[1,2-a]quinoxalin-5-one hydrochloride
Molecular Weight (g/mol):
308.59
Molecular Formula:
C11H12Cl3N3O
InChi Key:
YPNWSZJDAKOUAW-UHFFFAOYNA-N
SMILES:
Cl.ClC1=C(Cl)C=C2N3CCNCC3C(=O)NC2=C1
1
–
1
of
1
results
1
R&D Systems™ WAY 161503 hydrochloride
CAS: 276695-22-8 Molecular Formula: C11H12Cl3N3O