CAS RN 2767427-87-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ STING Degrader N-Me-SP23
Negative control for STING Degrader SP23 (Cat. No. 8053)