accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27816-36-0

IUPAC Name: 2-chloropropanamide
Synonyms: 2-chloropropanamide
Molecular Weight (g/mol): 107.54
Molecular Formula: C3H6ClNO
InChi Key: OEZPDHRXGCLGKB-UHFFFAOYNA-N
SMILES: CC(Cl)C(N)=O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results