accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOSOONHNOSOOO(E)

CAS RN 2783-94-0

IUPAC Name: disodium (5E)-6-oxo-5-[2-(4-sulfonatophenyl)hydrazin-1-ylidene]-5,6-dihydronaphthalene-2-sulfonate
Synonyms: sunset yellow fcf
Molecular Weight (g/mol): 452.36
Molecular Formula: C16H10N2Na2O7S2
InChi Key: TXVRKNUZLYFDTJ-DDVLFWKVSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(N\N=C2\C(=O)C=CC3=CC(=CC=C23)S([O-])(=O)=O)C=C1
Grade 
  • (1)
  • (1)
  • (4)
  • (1)
Color 
  • (4)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)