Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Food Yellow No. 5, TCI America™
SDP

Catalog No. F014525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F014525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F014525G Supplier TCI America Supplier No. F014525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity powder intended for laboratory research purposes.

  • Chemical Name: Food Yellow No. 5
  • Molecular Formula: C16H10N2Na2O7S2
  • Molecular Weight: 452.36 g/mol
  • Physical State: Powder
  • Color: Yellow red to deep yellow red
  • Solubility: Soluble in water (19 g/100 mL, 190 g/L) and glycerol; slightly soluble in propylene glycol; very slightly soluble in ethanol
  • Storage: Room temperature (recommended in a cool and dark place, <15°C)
  • Safety and Regulatory: IARC Group 3 (not classifiable as carcinogenic to humans); TSCA inventory listed; DSL listed

Chemical Identifiers

CAS 2783-94-0
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
MDL Number MFCD00036437
InChI Key TXVRKNUZLYFDTJ-UHFFFAOYSA-L
PubChem CID 6850717
IUPAC Name disodium;6-oxo-5-[(4-sulfonatophenyl)hydrazinylidene]naphthalene-2-sulfonate
SMILES C1=CC(=CC=C1NN=C2C3=C(C=CC2=O)C=C(C=C3)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Yellow
Quantity 25 g
Physical Form Powder
Formula Weight 452.36
Grade Research
Chemical Name or Material Food Yellow No. 5
EINECSNumber (5)-1451
RTECSNumber QK2450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.