accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2786-31-4

IUPAC Name: 3-methyl-1,3-benzothiazol-3-ium iodide
Synonyms: 3-methyl-1,3-benzothiazol-3-ium iodide
Molecular Weight (g/mol): 277.12
Molecular Formula: C8H8INS
InChi Key: SGYIRNXZLWJMCR-UHFFFAOYSA-M
SMILES: [I-].C[N+]1=CSC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results