accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3ONOHOOClClCl(E)(?)

CAS RN 278779-30-9

IUPAC Name: 3-[(1E)-2-(2-chloro-4-{[3-(2,6-dichlorophenyl)-5-(propan-2-yl)-1,2-oxazol-4-yl]methoxy}phenyl)ethenyl]benzoic acid
Synonyms: 3-[(1E)-2-(2-chloro-4-{[3-(2,6-dichlorophenyl)-5-isopropyl-1,2-oxazol-4-yl]methoxy}phenyl)ethenyl]benzoic acid
Molecular Weight (g/mol): 542.84
Molecular Formula: C28H22Cl3NO4
InChi Key: BYTNEISLBIENSA-MDZDMXLPSA-N
SMILES: CC(C)C1=C(COC2=CC=C(\C=C\C3=CC=CC(=C3)C(O)=O)C(Cl)=C2)C(=NO1)C1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)