accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2791-84-6

IUPAC Name: 1,5-dibenzyl 2-aminopentanedioate; 4-methylbenzene-1-sulfonic acid
Synonyms: 1,5-dibenzyl 2-aminopentanedioate; para-toluene sulfonate
Molecular Weight (g/mol): 499.58
Molecular Formula: C26H29NO7S
InChi Key: HVZUAIVKRYGQRM-UHFFFAOYNA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.NC(CCC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)