accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27969-56-8

IUPAC Name: 4-[(4Z)-3-methyl-4-[(2E,4E)-5-[5-methyl-3-oxo-2-(4-sulfophenyl)-2,3-dihydro-1H-pyrazol-4-yl]penta-2,4-dien-1-ylidene]-5-oxo-4,5-dihydro-1H-pyrazol-1-yl]benzene-1-sulfonic acid
Synonyms: 4-[(4Z)-3-methyl-4-[(2E,4E)-5-[3-methyl-5-oxo-1-(4-sulfophenyl)-2H-pyrazol-4-yl]penta-2,4-dien-1-ylidene]-5-oxopyrazol-1-yl]benzenesulfonic acid
Molecular Weight (g/mol): 570.59
Molecular Formula: C25H22N4O8S2
InChi Key: TZTOFZIBLYUIGG-GSLHRLMQSA-N
SMILES: CC1=C(\C=C\C=C\C=C2\C(C)=NN(C2=O)C2=CC=C(C=C2)S(O)(=O)=O)C(=O)N(N1)C1=CC=C(C=C1)S(O)(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results