accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 280-64-8

IUPAC Name: 9-borabicyclo[3.3.1]nonan-9-yl
Synonyms: 9-borabicyclo(3.3.1)nonane
Molecular Weight (g/mol): 121.01
Molecular Formula: C8H14B
InChi Key: AMKGKYQBASDDJB-UHFFFAOYSA-N
SMILES: [B]1C2CCCC1CCC2
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results