Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-BBN monomer, 97%, 0.5M in THF, Thermo Scientific™

Catalog No. AAH52287AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52287AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AAH52287AP Supplier Thermo Scientific Chemicals Supplier No. H52287AP

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
9-BBN monomer acts as a selective hydroboration reagent in synthetic organic chemistry. It is employed in Suzuki reactions as well as in the preparation of terminal alcohols by the region selective addition of alkenes followed by oxidative cleavage using hydrogen peroxide. It is also used in copper-catalyzed cross-coupling reactions of organoboron compounds with primary alkyl halides and pseudohalides. It acts as a protecting group for alkenes. Further, it is used as a reactant for Hetero-Diels-Alder reaction for the synthesis of spirocyclic alkaloids. In addition to this, it is used in intramolecular insertion of alkenes into palladium-nitrogen bonds.

Solubility
Miscible with ether, hexane, benzene, toluene, carbon tetrachloride, chloroform, dimethylsulfide and dichloromethane. Slightly miscible with cyclohexane, dimethoxyethane, diglyme and dioxane.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, oxidizing agents, acids and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 280-64-8
Molecular Formula C8H14B
Molecular Weight (g/mol) 121.01
MDL Number MFCD00074742
InChI Key AMKGKYQBASDDJB-UHFFFAOYSA-N
Synonym 9-borabicyclo 3.3.1 nonane, 9-borabicyclo 3.3.1 nonan, 9-borabicyclo 3,3,1 nonane, 9$l^ 2-borabicyclo 3.3.1 nonane, 1s,5s-9-borabicyclo 3.3.1 nonane, monomer; 21205-91-4 dimer, 9-borabicyclo 3.3.1 nonane,9-bbn,intetrahydrofuran
PubChem CID 6327450
IUPAC Name 9$l^{2}-borabicyclo[3.3.1]nonane
SMILES [B]1C2CCCC1CCC2

Specifications

Flash Point −19°C (−2°F)
Odor Ether-like
Quantity 500 mL
UN Number UN3399
Beilstein 605509
Sensitivity Air and moisture sensitive
Solubility Information Miscible with ether,hexane,benzene,toluene,carbon tetrachloride,chloroform,dimethylsulfide and dichloromethane. Slightly miscible with cyclohexane,dimethoxyethane,diglyme and dioxane.
Formula Weight 122.02
Percent Purity 97%
Concentration or Composition (by Analyte or Components) 0.5M in THF
Chemical Name or Material 9-BBN monomer
Show More Show Less
Hazard Category H225-H261-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H351-H319-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Ambient temperatures; Store under Argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.