accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 280560-78-3

IUPAC Name: 1-[(4-bromophenyl)methyl]-4-methyl-1,4-diazepane
Synonyms: 1-[(4-bromophenyl)methyl]-4-methyl-1,4-diazepane
Molecular Weight (g/mol): 283.21
Molecular Formula: C13H19BrN2
InChi Key: WQUWRVLHCKJAMG-UHFFFAOYSA-N
SMILES: CN1CCCN(CC2=CC=C(Br)C=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results