accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28069-72-9

IUPAC Name: (2E,6E)-nona-2,6-dien-1-ol
Synonyms: nonadienol
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: AMXYRHBJZOVHOL-DYWGDJMRSA-N
SMILES: CC\C=C\CC\C=C\CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results