accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNONHOClCl

CAS RN 280744-09-4

IUPAC Name: 3-(2,4-dichlorophenyl)-4-(1-methyl-1H-indol-3-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 3-(2,4-dichlorophenyl)-4-(1-methylindol-3-yl)-1H-pyrrole-2,5-dione
Molecular Weight (g/mol): 371.22
Molecular Formula: C19H12Cl2N2O2
InChi Key: JCSGFHVFHSKIJH-UHFFFAOYSA-N
SMILES: CN1C=C(C2=CC=CC=C12)C1=C(C(=O)NC1=O)C1=CC=C(Cl)C=C1Cl
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results