accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28163-00-0

IUPAC Name: 2-chloro-4-nitrobenzonitrile
Synonyms: 2-chloro-4-nitrobenzonitrile
Molecular Weight (g/mol): 182.56
Molecular Formula: C7H3ClN2O2
InChi Key: ZIGQFRQZYVQQDR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C#N)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results