CAS RN 2821803-61-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ HEMTAC 26
CDK6 and CDK4 HSP90-mediated targeting chimera (HEMTAC) Degrader