accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 28257-92-3

CH3H3COHHOOOHOOH

CAS RN 28257-92-3

IUPAC Name: 2,2-dimethylpropane-1,3-diol; butanedioic acid
Synonyms: neopentyl glycol; succinic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C9H18O6
InChi Key: GXTJJXHLTYJRLX-UHFFFAOYSA-N
SMILES: CC(C)(CO)CO.OC(=O)CCC(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results