CAS RN 28322-40-9
IUPAC Name:
(3-methylbutyl)triphenylphosphanium bromide
Synonyms:
(3-methylbutyl)triphenylphosphanium bromide
Molecular Weight (g/mol):
413.34
Molecular Formula:
C23H26BrP
InChi Key:
GZLGTVRDLCJQTO-UHFFFAOYSA-M
SMILES:
[Br-].CC(C)CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich Phenol-1,2-Dichlorobenzene Mixture
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.