CAS RN 2835-81-6
DL-2-Aminobutyric acid, 99%
CAS: 2835-81-6 Molecular Formula: C4H9NO2 Molecular Weight (g/mol): 103.121 MDL Number: MFCD00008093 InChI Key: QWCKQJZIFLGMSD-UHFFFAOYSA-N Synonym: dl-2-aminobutyric acid,2-aminobutyric acid,alpha-aminobutyric acid,butanoic acid, 2-amino,butyrine,dl-2-amino-n-butyric acid,h-dl-abu-oh,aaba,dl-alpha-amino-n-butyric acid,alpha-amino-n-butyric acid PubChem CID: 6657 ChEBI: CHEBI:35621 IUPAC Name: 2-aminobutanoic acid SMILES: CCC(C(=O)O)N
DL-2-Aminobutyric acid, 99%
CAS: 2835-81-6 Molecular Formula: C4H9NO2 Molecular Weight (g/mol): 103.12 InChI Key: QWCKQJZIFLGMSD-UHFFFAOYSA-N Synonym: dl-2-aminobutyric acid,2-aminobutyric acid,alpha-aminobutyric acid,butanoic acid, 2-amino,butyrine,dl-2-amino-n-butyric acid,h-dl-abu-oh,aaba,dl-alpha-amino-n-butyric acid,alpha-amino-n-butyric acid PubChem CID: 6657 ChEBI: CHEBI:35621 IUPAC Name: 2-aminobutanoic acid SMILES: CCC(C(=O)O)N
DL-2-Aminobutyric Acid 99.0+%, TCI America™
CAS: 2835-81-6 Molecular Formula: C4H9NO2 Molecular Weight (g/mol): 103.121 MDL Number: MFCD00008093 InChI Key: QWCKQJZIFLGMSD-UHFFFAOYSA-N Synonym: dl-2-aminobutyric acid,2-aminobutyric acid,alpha-aminobutyric acid,butanoic acid, 2-amino,butyrine,dl-2-amino-n-butyric acid,h-dl-abu-oh,aaba,dl-alpha-amino-n-butyric acid,alpha-amino-n-butyric acid PubChem CID: 6657 ChEBI: CHEBI:35621 IUPAC Name: 2-aminobutanoic acid SMILES: CCC(C(=O)O)N
Sigma Aldrich Dl-2-Aminobutyric Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich (S)-1-Boc-4-Oxopiperidine-2-Carboxylic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc DL-α-Amino-n-butyric Acid, MP Biomedicals
CAS: 2835-81-6 Molecular Formula: C4H9NO2 Molecular Weight (g/mol): 103.121 InChI Key: QWCKQJZIFLGMSD-UHFFFAOYSA-N Synonym: dl-2-aminobutyric acid,2-aminobutyric acid,alpha-aminobutyric acid,butanoic acid, 2-amino,butyrine,dl-2-amino-n-butyric acid,h-dl-abu-oh,aaba,dl-alpha-amino-n-butyric acid,alpha-amino-n-butyric acid PubChem CID: 6657 ChEBI: CHEBI:35621 IUPAC Name: 2-aminobutanoic acid SMILES: CCC(C(=O)O)N