Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-2-Aminobutyric acid, 99%

Catalog No. AC172091000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172091000 100 g Glass bottle
AC172090250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172091000 Supplier Thermo Scientific Chemicals Supplier No. 172091000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2835-81-6 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2835-81-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
InChI Key QWCKQJZIFLGMSD-UHFFFAOYSA-N
Synonym dl-2-aminobutyric acid, 2-aminobutyric acid, alpha-aminobutyric acid, butanoic acid, 2-amino, butyrine, dl-2-amino-n-butyric acid, h-dl-abu-oh, aaba, dl-alpha-amino-n-butyric acid, alpha-amino-n-butyric acid
PubChem CID 6657
ChEBI CHEBI:35621
IUPAC Name 2-aminobutanoic acid
SMILES CCC(C(=O)O)N

Specifications

Melting Point 291.0°C to 293.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C2H5CH(NH2)CO2H
Quantity 100 g
Beilstein 04,408
Merck Index 15,423
Solubility Information Solubility in water: 210g/L (25 c). Other solubilities: sparingly soluble in alcohol,insoluble in ether
Formula Weight 103.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material DL-2-Aminobutyric acid, 99+%
Show More Show Less
EINECSNumber 220-616-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.