Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-α-Amino-n-butyric Acid, MP Biomedicals

Catalog No. ICN207535 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN207535 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN207535 Supplier MP Biomedicals, Inc Supplier No. 0520753510

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2835-81-6
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
InChI Key QWCKQJZIFLGMSD-UHFFFAOYSA-N
Synonym dl-2-aminobutyric acid, 2-aminobutyric acid, alpha-aminobutyric acid, butanoic acid, 2-amino, butyrine, dl-2-amino-n-butyric acid, h-dl-abu-oh, aaba, dl-alpha-amino-n-butyric acid, alpha-amino-n-butyric acid
PubChem CID 6657
ChEBI CHEBI:35621
IUPAC Name 2-aminobutanoic acid
SMILES CCC(C(=O)O)N

Specifications

Quantity 10 g
Formula Weight 103.12
Chemical Name or Material DL-α-Amino-n-butyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.