accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH

CAS RN 28393-06-8

IUPAC Name: heptadeca-10,12-diynoic acid
Synonyms: heptadeca-10,12-diynoic acid
Molecular Weight (g/mol): 262.39
Molecular Formula: C17H26O2
InChi Key: CLTRJQSBWDSZHN-UHFFFAOYSA-N
SMILES: CCCCC#CC#CCCCCCCCCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results