CAS RN 2839486-16-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ CBMR 0083
Potent allosteric FFA1 (GPR40) and 4 (GPR120) agonist