accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONOO

CAS RN 284679-97-6

IUPAC Name: 3-nitro-4-(pyrrolidin-1-yl)benzaldehyde
Synonyms: 3-nitro-4-(pyrrolidin-1-yl)benzaldehyde
Molecular Weight (g/mol): 220.23
Molecular Formula: C11H12N2O3
InChi Key: AONHBESCCBXPAD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(C=O)=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results