accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOFFFFFF

CAS RN 28523-86-6

IUPAC Name: 1,1,1,3,3,3-hexafluoro-2-(fluoromethoxy)propane
Synonyms: sevoflurane sojourn sevoflurane product sevoflurane (substance) sevoflurano sevoflurane (product) sevorane sevoflurano [inn-spanish] fluoromethyl hexafluoroisopropyl ether sevofluranum
Molecular Weight (g/mol): 200.06
Molecular Formula: C4H3F7O
InChi Key: DFEYYRMXOJXZRJ-UHFFFAOYSA-N
SMILES: FCOC(C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)