Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoromethyl 1,1,1,3,3,3-Hexafluoroisopropyl Ether 98.0+%, TCI America™
SDP

Catalog No. F06915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F06915G Supplier TCI America Supplier No. F06915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28523-86-6
Molecular Formula C4H3F7O
Molecular Weight (g/mol) 200.056
MDL Number MFCD00153189
InChI Key DFEYYRMXOJXZRJ-UHFFFAOYSA-N
Synonym sevoflurane, sevofluran, ultane, 1,1,1,3,3,3-hexafluoro-2-fluoromethoxy propane, sevoflurano, sevofluranum, sevorane, sojourn, mr6s4, sevofluranum inn-latin
PubChem CID 5206
ChEBI CHEBI:9130
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-(fluoromethoxy)propane
SMILES C(OC(C(F)(F)F)C(F)(F)F)F

Specifications

Color Colorless
Boiling Point 58°C
Quantity 5 g
Formula Weight 200.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Fluoromethyl 1,1,1,3,3,3-Hexafluoroisopropyl Ether
RTECSNumber KO0737000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.