accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONHOHOOClHNOOHONHOOOHHNOCH3

CAS RN 285570-60-7

IUPAC Name: 3-{2-[4-carboxy-2-(3-carboxy-2-acetamidopropanamido)butanamido]-3-methylbutanamido}-5-chloro-4-oxopentanoic acid
Synonyms: 3-{2-[4-carboxy-2-(3-carboxy-2-acetamidopropanamido)butanamido]-3-methylbutanamido}-5-chloro-4-oxopentanoic acid
Molecular Weight (g/mol): 550.95
Molecular Formula: C21H31ClN4O11
InChi Key: ATNOUPFYBMVFLD-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)C(CCC(O)=O)NC(=O)C(CC(O)=O)NC(C)=O)C(=O)NC(CC(O)=O)C(=O)CCl
Form 
  • (2)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results