accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CHO

CAS RN 28664-35-9

IUPAC Name: 3-hydroxy-4,5-dimethyl-2,5-dihydrofuran-2-one
Synonyms: sotolon
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: UNYNVICDCJHOPO-UHFFFAOYNA-N
SMILES: CC1OC(=O)C(O)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results