Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethyl-3-hydroxy-2(5H)-furanone (ca. 14% in Propylene Glycol, ca. 1.2mol/L), TCI America™
SDP

Catalog No. D18515ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18515ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D18515ML Supplier TCI America Supplier No. D18515ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28664-35-9
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.127
MDL Number MFCD00059957
InChI Key UNYNVICDCJHOPO-UHFFFAOYSA-N
Synonym sotolone, 3-hydroxy-4,5-dimethylfuran-2 5h-one, sotolon, 4,5-dimethyl-3-hydroxy-2,5-dihydrofuran-2-one, 4,5-dimethyl-3-hydroxy-2 5h-furanone, 2 5h-furanone, 3-hydroxy-4,5-dimethyl, 3-hydroxy-4,5-dimethyl-2 5h-furanone, caramel furanone, fenugreek lactone, 2-hydroxy-3-methyl-2-penten-4-olide
PubChem CID 62835
ChEBI CHEBI:67890
IUPAC Name 4-hydroxy-2,3-dimethyl-2H-furan-5-one
SMILES CC1C(=C(C(=O)O1)O)C

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 128.13
Physical Form Liquid
Chemical Name or Material 4,5-Dimethyl-3-hydroxy-2(5H)-furanone (ca. 14% in Propylene Glycol, ca. 1.2mol/L)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.