CAS RN 2866429-93-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Disulfide biotin azide
Azide-activated biotinylation reagent; labels alkyne moieties via copper-catalyzed click chemistry