accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNH2OOHOOHOOOHOHOHNHOH3CNHOCH3

CAS RN 286959-52-2

IUPAC Name: 2-amino-3-[(3-acetamido-4-{[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-5-hydroxy-6-(hydroxymethyl)oxan-2-yl)oxy]butanoic acid
Synonyms: 2-amino-3-[(3-acetamido-4-{[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-5-hydroxy-6-(hydroxymethyl)oxan-2-yl)oxy]butanoic acid
Molecular Weight (g/mol): 525.51
Molecular Formula: C20H35N3O13
InChi Key: BMSPZGKRTLZXAV-UHFFFAOYNA-N
SMILES: CC(OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2NC(C)=O)C1NC(C)=O)C(N)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results