accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOH(R)OH(S)OHO

CAS RN 28697-53-2

IUPAC Name: (2S,3R)-2,3,4,5-tetrahydroxypentanal
Synonyms: (2S,3R)-2,3,4,5-tetrahydroxypentanal
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: PYMYPHUHKUWMLA-DEOSMSJNSA-N
SMILES: OCC(O)[C@@H](O)[C@H](O)C=O
Quantity 
  • (1)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)