Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-Arabinose, ≥99%, MP Biomedicals™

Catalog No. ICN10069925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10069925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10069925 Supplier MP Biomedicals, Inc Supplier No. 0210069925

missing translation for 'validMainframeMsg'

D-(—)-Arabinose is a substrate used to identify, differentiate and characterize arabinose isomerase(s) that have commercial value in the production of tagatose, a low calorie sweetener

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28697-53-2
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
InChI Key PYMYPHUHKUWMLA-DEOSMSJNSA-N
Synonym alpha-d-arabinopyranose, 2s,3s,4r,5r-oxane-2,3,4,5-tetrol, d--arabinose, alpha-arabinose, a-d-arabinopyranose, alpha-d-arabinopyranose 9ci, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
PubChem CID 71313478
IUPAC Name (2S,3R)-2,3,4,5-tetrahydroxypentanal
SMILES OCC(O)[C@@H](O)[C@H](O)C=O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 25 g
Formula Weight 150.13
Physical Form Powder
Chemical Name or Material D-(-)-Arabinose
EINECSNumber 233-708-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.