accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOONHOHNSOOOSOOOOSOOOKKKK(E/Z)

CAS RN 28699-96-9

IUPAC Name: tetrapotassium 3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5',7,7'-tetrasulfonate
Synonyms: tetrapotassium 3,3'-dioxo-1H,1'H-[2,2'-biindolylidene]-5,5',7,7'-tetrasulfonate
Molecular Weight (g/mol): 734.86
Molecular Formula: C16H6K4N2O14S4
InChi Key: SWRJFVUVNXWCRX-UHFFFAOYSA-J
SMILES: [K+].[K+].[K+].[K+].[O-]S(=O)(=O)C1=CC(=C2NC(C(=O)C2=C1)=C1NC2=C(C=C(C=C2C1=O)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results