accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2873-90-7

IUPAC Name: 4-(diethylamino)benzonitrile
Synonyms: 4-(diethylamino)benzonitrile
Molecular Weight (g/mol): 174.25
Molecular Formula: C11H14N2
InChi Key: KMLGFOAKCYHXCQ-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results