CAS RN 287403-39-8
IUPAC Name:
4-[4-(but-2-yn-1-yloxy)benzenesulfonyl]-N-hydroxy-2,2-dimethylthiomorpholine-3-carboxamide
Synonyms:
4-[4-(but-2-yn-1-yloxy)benzenesulfonyl]-N-hydroxy-2,2-dimethylthiomorpholine-3-carboxamide
Molecular Weight (g/mol):
398.49
Molecular Formula:
C17H22N2O5S2
InChi Key:
CVZIHNYAZLXRRS-UHFFFAOYNA-N
SMILES:
CC#CCOC1=CC=C(C=C1)S(=O)(=O)N1CCSC(C)(C)C1C(=O)NO
1
–
2
of
2
results
1
R&D Systems™ TMI 1, >98%
CAS: 287403-39-8 Molecular Formula: C17H22N2O5S2
2
R&D Systems™ SI-2 hydrochloride, >98%
CAS: 287403-39-8 Molecular Formula: C17H22N2O5S2