accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONN

CAS RN 2876-17-7

IUPAC Name: 1-methoxyphenazine
Synonyms: 1-methoxyphenazine
Molecular Weight (g/mol): 210.24
Molecular Formula: C13H10N2O
InChi Key: ZYDGCYWJDWIJCS-UHFFFAOYSA-N
SMILES: COC1=C2N=C3C=CC=CC3=NC2=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results