Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methoxyphenazine 97.0+%, TCI America™
SDP

Catalog No. M18281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18281G Supplier TCI America Supplier No. M18281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2876-17-7
Molecular Formula C13H10N2O
Molecular Weight (g/mol) 210.236
MDL Number MFCD01794656
InChI Key ZYDGCYWJDWIJCS-UHFFFAOYSA-N
PubChem CID 76137
IUPAC Name 1-methoxyphenazine
SMILES COC1=CC=CC2=NC3=CC=CC=C3N=C21

Specifications

Melting Point 171°C
Color Yellow
Quantity 1 g
Formula Weight 210.24
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methoxyphenazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.